Merck
  • [Influence of occlusal contacts on tooth displacement for mesio-distal direction].
[Influence of occlusal contacts on tooth displacement for mesio-distal direction].
Kokubyo Gakkai zasshi. The Journal of the Stomatological Society, Japan (2004-04-24)
Ayako Tokuda
ABSTRACT
To fabricate a prosthesis which is functionally harmonized with the stomatognathic system, the interocclusal relationship must be reproduced as accurately as possible. This study investigated the influence of the occlusal contacts upon tooth displacement and clarified the optimal occlusal contacts for the prosthesis. The displacement paths of six maxillary left first molars of six adults in good general health were measured in this study. We gave each test tooth eight patterns of artificial occlusal contacts by attaching platinum foil on the occlusal surface. Subjects were asked to perform clenching at the intercuspal position (= ICP) with medium occlusal forces, and 3-dimensional tooth displacement paths were measured by the Type M-3 tooth displacement transducer. The following conclusions were obtained. 1. Using the Student's t-test for statistical analysis, it was suggested that the difference of the buccolingual location of occlusal contacts influenced the direction of the tooth displacement. 2. The tooth which had occlusal contact in the buccal cusp displaced in the mesio-buccal direction, so this may create a wide dental arch and a slight discrepancy was generated at the distal interproximal contact area of the tooth. 3. It was shown that the tooth which had occlusal contact in the buccal cusp rotated in the buccoapical direction in reverse to the natural tooth.
Product Description
Platinum, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.002mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.25mm, as rolled, 99.999%
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platinum, foil, not light tested, 100x100mm, thickness 0.0075mm, as rolled, 99.95%
Platinum, foil, 0.1m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.0075mm, as rolled, 99.95%
Platinum, foil, 0.1m coil, thickness 0.2mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, 0.05m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Platinum, foil, 10mm disks, thickness 0.01mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.0125mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platinum, foil, 6mm disks, thickness 0.0075mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.125mm, coil width 2.5mm, as rolled, 99.99%
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width 1.2mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.030mm, coil width .7mm, as rolled, 99.95%
Platinum, foil, 0.1m coil, thickness 0.025mm, coil width 8mm, as rolled, 99.95%
Platinum, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95%
Platinum, foil, not light tested, 100x100mm, thickness 0.004mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Platinum, foil, 4mm disks, thickness 0.1mm, as rolled, 99.99+%
Platinum, foil, 15mm disks, thickness 0.1mm, as rolled, 99.99+%
Platinum, foil, 4mm disks, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, not light tested, 100x100mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, 10mm disks, thickness 0.05mm, as rolled, 99.95%
Platinum, foil, 6mm disks, thickness 0.025mm, as rolled, 99.95%