Merck
A review of tungsten: from environmental obscurity to scrutiny.
Journal of hazardous materials (2005-12-14)
A Koutsospyros, W Braida, C Christodoulatos, D Dermatas, N Strigul
ABSTRACT
Since its discovery, tungsten, a transition element of Group VIb of the Periodic Table of Elements, and its compounds have been considered environmentally benign. Its presence in biological and drinking water samples in Fallon, Nevada, an acute lymphocytic leukemia cluster struck community has alarmed public health, environmental and regulatory agencies. Tungsten, a metal of extraordinary properties that make it hardly substitutable, is considered an essential commodity with a wide variety of uses stretching from household necessities to highly specialized applications. This work is undertaken in order to explore aspects of environmental behavior of tungsten and its compounds. Occurrence data in terrestrial, atmospheric, aquatic and biotic systems are presented. Various aspects of environmental chemistry, fate transport across environmental interfaces and toxicology are discussed with the objective of identifying knowledge gaps and outlining directions for future research.
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Tungsten, insulated wire, 5m, conductor diameter 0.075mm, insulation thickness 0.02mm, PTFE (polytetrafluoroethylene) insulation, 99.9%
Tungsten, insulated wire, 10m, conductor diameter 0.05mm, insulation thickness 0.008mm, polyimide insulation, 99.9%
Tungsten, microfoil, disks, 10mm, thinness 0.25μm, specific density 474μg/cm2, permanent mylar 3.5μm support, 99.95%
Tungsten, insulated wire, 0.2m, conductor diameter 0.015mm, insulation thickness 0.002mm, polyimide insulation, 99.9%
Tungsten, foil, 5m coil, thickness 0.025mm, coil width 1mm, as rolled, 99.95%
Tungsten, insulated wire, 20m, conductor diameter 0.05mm, insulation thickness 0.008mm, polyimide insulation, 99.9%
Tungsten, foil, not light tested, 50x50mm, thickness 0.004mm, 99.95%
Tungsten, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95%
Tungsten, lump, 200g, max. size 5mm
Tungsten, insulated wire, 2m, conductor diameter 0.075mm, insulation thickness 0.02mm, PTFE (polytetrafluoroethylene) insulation, 99.9%
Tungsten, rod, 200mm, diameter 6.35mm, centerless ground, 99.95%
Tungsten, foil, light tested, 50x50mm, thickness 0.075mm, as rolled, 99.95%
Tungsten, foil, not light tested, 25x25mm, thickness 0.004mm, 99.95%
Tungsten, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyimide insulation, 99.9%
Tungsten, foil, 30mm disks, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, light tested, 150x150mm, thickness 0.075mm, as rolled, 99.95%
Tungsten, lump, 50g, max. size 5mm
Tungsten, insulated wire, 1m, conductor diameter 0.015mm, insulation thickness 0.002mm, polyimide insulation, 99.9%
Tungsten, microfoil, disks, 10mm, thinness 0.05μm, specific density 96μg/cm2, permanent mylar 3.5μm support, 99.95%
Tungsten, insulated wire, 2m, conductor diameter 0.0125mm, insulation thickness 0.0035mm, polyimide insulation, 99.9%
Tungsten, foil, light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95%
Tungsten, microfoil, disks, 10mm, thinness 0.1μm, specific density 182μg/cm2, permanent mylar 3.5μm support, 99.95%
Tungsten, wire reel, 0.5m, diameter 1.0mm, clean, 99.95%
Tungsten, insulated wire, 2m, conductor diameter 0.015mm, insulation thickness 0.002mm, polyimide insulation, 99.9%
Tungsten, foil, not light tested, 50x50mm, thickness 0.007mm, 99.95%
Tungsten, foil, not light tested, 25x25mm, thickness 0.006mm, 99.95%
Tungsten, rod, 100mm, diameter 5.0mm, centerless ground, 99.95%
Tungsten, mesh, 100x100mm, nominal aperture 0.2mm, wire diameter 0.05mm, 100 wires/inch, open area 64%, plain weave mesh, 99.95%