Merck
  • [Levitra (oral dispersible tablet)--an innovative drug for the treatment of patients with erectile dysfunction].
[Levitra (oral dispersible tablet)--an innovative drug for the treatment of patients with erectile dysfunction].
Urologiia (Moscow, Russia : 1999) (2013-06-26)
I A Korneev
ABSTRACT
The arsenal of professionals providing assistance to men with erectile dysfunction includes a new, mouth dissolving dosage form of the vardenafil--levitra ODT. This form is effective, safe, and has a higher bioavailability as compared to vardenafil in the form of coated tablets. POTENT I and POTENT II studies revealed that levitra ODT equally successfully corrects erectile dysfunction in men younger and older than 65 years regardless of concomitant diseases and metabolic disorders: hypertension, diabetes, dyslipidemia. In addition, it was found that this drug helped to successfully carry out sexual intercourse in the first 15 minutes after dosing in 62.5% of patients. This allows to widely recommend levitra ODT for the treatment of erectile dysfunction in young men and older men.
Product Description
Supelco
Zopiclone-D4 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Vardenafil dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®