Merck
  • Sphingomyelin metabolism at the plasma membrane: implications for bioactive sphingolipids.
Sphingomyelin metabolism at the plasma membrane: implications for bioactive sphingolipids.
FEBS letters (2009-10-28)
Delphine Milhas, Christopher J Clarke, Yusuf A Hannun
ABSTRACT
The plasma membrane (PM) is a major resource for production of bioactive lipids and contains a large proportion of the cellular sphingomyelin (SM) content. Consequently, the regulation of SM levels at the PM by enzymes such as sphingomyelinase (SMase) and SM synthase 2 (SMS2) can have profound effects - both on biophysical properties of the membrane, but also on cellular signaling. Over the past 20 years, there has been considerable research into the physiological and cellular functions associated with regulation of SM levels, notably with regards to the production of ceramide. In this review, we will summarize this research with particular focus on the SMases and SMS2. We will outline what biological functions are associated with SM metabolism/production at the PM, and discuss what we believe are major challenges that need to be addressed in future studies.
Product Description
Avanti
Brain SM, Avanti Research - A Croda Brand
Avanti
18:1 SM (d18:1/18:1(9Z)), Avanti Research - A Croda Brand 860587P, powder
Avanti
Brain SM, Avanti Research - A Croda Brand
Avanti
Sphingomyelin (d18:1/12:0), Avanti Research - A Croda Brand LM2312, ethanol solution
Avanti
17:0 SM (d18:1/17:0), Avanti Research - A Croda Brand 860585P, powder
Avanti
18:1 SM (d18:1/18:1(9Z)), Avanti Research - A Croda Brand 860587C
Avanti
18:0 SM (d18:1/18:0), Avanti Research - A Croda Brand 860586P, powder
Avanti
06:0 SM (d18:1/6:0), Avanti Research - A Croda Brand 860582C
Avanti
02:0 SM (d18:1/2:0), Avanti Research - A Croda Brand 860580P, powder
Avanti
24:1 SM, Avanti Research - A Croda Brand, powder
Avanti
16:0 SM (d18:1/16:0), Avanti Research - A Croda Brand, powder
Avanti
24:0 SM, Avanti Research - A Croda Brand 860592P, powder
Avanti
Milk SM, Avanti Research - A Croda Brand
Avanti
Egg SM, Avanti Research - A Croda Brand
Avanti
Milk SM, Sphingomyelin (Milk, Bovine), chloroform
Avanti
17:0-20:4 PI(3,4,5)P3, Avanti Research - A Croda Brand LM1906, powder
Avanti
Egg SM, Avanti Research - A Croda Brand
Avanti
12:0 SM (d18:1/12:0), Avanti Research - A Croda Brand 860583P, powder
Avanti
06:0 SM (d18:1/6:0), Avanti Research - A Croda Brand 860582P, powder