Merck
  • Chemical mutagenesis testing in Drosophila. IX. Results of 50 coded compounds tested for the National Toxicology Program.
Chemical mutagenesis testing in Drosophila. IX. Results of 50 coded compounds tested for the National Toxicology Program.
Environmental and molecular mutagenesis (1994-01-01)
P Foureman, J M Mason, R Valencia, S Zimmering
ABSTRACT
Fifty chemicals were tested for mutagenic activity in post-meiotic and meiotic germ cells of male Drosophila melanogaster using the sex-linked recessive lethal (SLRL) assay. As in the previous studies in this series, feeding was chosen as the first route of administration. If the compound failed to induce mutations by this route, injection exposure was used. One gaseous chemical (1,3-butadiene) was tested only by inhalation. Those chemicals that were mutagenic in the sex-linked recessive lethal assay were further tested for the ability to induce reciprocal translocations. Eleven of the 50 chemicals tested were mutagenic in the SLRL assay. These included bis(2-chloroethyl) ether, 1,4-butanediol diglycidyl ether, 1-chloro-2-propanol, dimethyl methylphosphonate, dimethyl morpholinophosphoramidate, dimethyloldihydroxyethylene urea, 2,2-dimethyl vinyl chloride, hexamethylphosphoramide, isatin-5-sulfonic acid (Na salt), isopropyl glycidyl ether, and urethane. Five of these, including 1,4-butanediol diglycidyl ether, 2,2-dimethyl vinyl chloride, hexamethylphosphoramide, isopropyl glycidyl ether, and urethane, also induced reciprocal translocations.
Sigma-Aldrich
2-Chloroethyl ether, 99%
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, ≥95%
Sigma-Aldrich
Bis(2-chloroethyl) ether, puriss., ≥99.0% (GC)
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, technical grade, 60%
Sigma-Aldrich
1-Chloro-2-propanol, 70%
Sigma-Aldrich
Glycidyl isopropyl ether, 98%