Merck
  • ZIO impregnation and cytochemical localization of thiamine pyrophosphatase and acid phosphatase activities in small granule-containing (SGC) cells of rat superior cervical ganglia.
ZIO impregnation and cytochemical localization of thiamine pyrophosphatase and acid phosphatase activities in small granule-containing (SGC) cells of rat superior cervical ganglia.
Histology and histopathology (1994-10-01)
Y P Chau, K S Lu
ABSTRACT
Cytochemical relationship between Golgi complex and dense-cored granules (DCGs) of small granule-containing (SGC) cells in rat superior cervical ganglia was examined in electron microscopy by zinc-iodide-osmium tetroxide (ZIO) method and by enzyme cytochemistry for thiamine pyrophosphatase (TPPase) and acid phosphatase (ACPase). After ZIO impregnation, all the saccules of Golgi apparatus and some of tubular rough endoplasmic reticulum (rER) were stained. DCGs in periphery of SGC cells were not stained, but varying degrees of dense deposits occurred in the DCGs in vicinity of Golgi trans-saccules. Both TPPase and ACPase activities were localized in one or two stacked layers of saccules on the trans side of the Golgi complex. No reaction products were demonstrated in the DCGs. From these results, we suggest that the DCGs of SGC cells in rat superior cervical ganglia are derived from the Golgi complex, and that lysosomal cleavage of protein contents in the DCGs may occur in the trans Golgi saccules.
Sigma-Aldrich
Zinc iodide, ≥98%
Sigma-Aldrich
Zinc iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Zinc iodide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Zinc iodide, purum p.a., ≥98.0% (AT)