Merck
  • Vitamin K4 induces tumor cytotoxicity in human prostate carcinoma PC-3 cells via the mitochondria-related apoptotic pathway.
Vitamin K4 induces tumor cytotoxicity in human prostate carcinoma PC-3 cells via the mitochondria-related apoptotic pathway.
Die Pharmazie (2013-07-24)
Yu Jiang, Jingbo Yang, Chunhui Yang, Fanmin Meng, Yongfeng Zhou, Bo Yu, Muhammad Khan, Hong Yang
ABSTRACT
Vitamin K4 (VK4) is a synthetic hydrophilic menadione compound, which is clinically used as hemostasis medicine. It has been reported that several vitamin Ks had inhibitory effects on various cancer cells. However, there is no report about VK4s anticancer activity. The goal of this study was to investigate the inhibitory effect of VK4 on human prostate PC-3 cells and the mechanisms involved. We found that VK4 dose-dependently inhibited cell proliferation in PC-3 cells with an IC50 value of about 20.94 microM. Hoechst 33258 Staining results showed that VK4 caused DNA fragmentation in PC-3 cells. PI staining results indicated that VK4-induced PC-3 cell cycle arrest at the S phase. Further mechanistic studies revealed that VK4-mediated induction of apoptosis in PC-3 cell is associated with disruption of mitochondrial membrane potential, down-regulation of Bcl-2, and up-regulation of Bax, release of cytochrome c from mitochondria, and activation of caspase-3 and PARR Thus, VK4 might be useful in prostate cancer chemotherapy.
Sigma-Aldrich
Menadione sodium bisulfite, BioReagent, suitable for cell culture, ≥95% (TLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Menadione sodium bisulfite, ≥95% (TLC)
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications
Supelco
Menadione (K3), analytical standard
Menadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vitamin K4, ≥98% (HPLC)