Merck
  • The effect of Helicobacter pylori on vitamin B 12 blood levels in chronic renal failure patients: a single blind control trial.
The effect of Helicobacter pylori on vitamin B 12 blood levels in chronic renal failure patients: a single blind control trial.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia (2013-07-03)
Hossein Khedmat, Mohsen Amini, Ashraf Karbasi, Reza Azizi
ABSTRACT
Helicobacter pylori (HP) is a common infection worldwide and has been associated with severe morbidity. The level of vitamin B 12 in HP-infected chronic kidney disease (CKD) patients is reported to be lower than in the general population. The present study has been designed to evaluate the vitamin B 12 level in HP-infected CKD patients. We assessed the serum levels of vitamin B 12 in 50 CKD patients with positive HP serology, one and three months after the eradication of HP infection. There were significant differences between the serum levels of vitamin B 12 in the study patients before (806.98 ± 466.82) and after (760.36 ± 433.93) eradication treatment (P <0.001). We conclude that our study suggests the correlation between vitamin B 12 deficiency in CKD patients and the HP infection status.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin (B12), analytical standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material