Merck
  • Chromium and cobalt ion concentrations in blood and serum following various types of metal-on-metal hip arthroplasties: a literature overview.
Chromium and cobalt ion concentrations in blood and serum following various types of metal-on-metal hip arthroplasties: a literature overview.
Acta orthopaedica (2013-04-19)
Christopher Jantzen, Henrik L Jørgensen, Benn R Duus, Sune L Sporring, Jes B Lauritzen
ABSTRACT
Widely different metal ion concentrations in blood and serum have been reported with metal-on-metal (MoM) implants. We reviewed the literature on blood and serum ion concentrations of chromium (Cr) and cobalt (Co) following various MoM hip arthroplasties. Studies were searched for in the Medline database, Embase, and the Cochrane Database of Systematic Reviews. Highest mean or median ion concentrations of Cr and Co after a minimum of 1 year of follow-up were extracted and grouped according to sample- and articulation type, and average values were calculated. 43 studies were included and 16 different MoM implants were identified. For the different types of bearings, average ion concentrations and range were calculated from the mean or median ion concentration. The average Cr concentration ranged between 0.5 and 2.5 μg/L in blood and between 0.8 and 5.1 μg/L in serum. For Co, the range was 0.7-3.4 μg/L in blood and 0.3-7.5 μg/L in serum. When the average blood ion concentrations calculated for the different implants, together with the concentrations measured in the individual studies, were compared with the upper acceptable limit for Cr and Co in blood, no clear pattern was recognized. Furthermore, we were unable to detect any clear difference in ion concentrations between different types of implants (THA and resurfacing).
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cobalt, foil, not light tested, 150x150mm, thickness 0.0125mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.02mm, 99.9%
Cobalt, wire reel, 0.1m, diameter 1.0mm, hard, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.007mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.0125mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Cobalt, foil, not light tested, 150x150mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.005mm, 99.9%