Merck
  • Postsurgical intrapericardial adhesions: mechanisms of formation and prevention.
Postsurgical intrapericardial adhesions: mechanisms of formation and prevention.
The Annals of thoracic surgery (2013-04-10)
Aldo Cannata, Duccio Petrella, Claudio Francesco Russo, Giuseppe Bruschi, Pasquale Fratto, Marcello Gambacorta, Luigi Martinelli
ABSTRACT
Postsurgical intrapericardial adhesions are still considered an unavoidable consequence of cardiothoracic operations. They increase the technical difficulty and the risk of reoperations. The pathogenesis of postsurgical adhesions is a multistep process, and the main key players are (1) loss of mesothelial cells, (2) accumulation of fibrin in areas devoid of mesothelial cells, (3) loss of normal pericardial fibrinolysis, and (4) local inflammation. Today, very promising methods to reduce adhesions are available for clinical use. This report reviews the process of formation of adhesions and the methods to prevent them, classified according to the mechanism of action.
Sigma-Aldrich
Polytetrafluoroethylene preparation, 60 wt % dispersion in H2O
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads