Merck
  • Clinical and laboratory findings in border collies with presumed hereditary juvenile cobalamin deficiency.
Clinical and laboratory findings in border collies with presumed hereditary juvenile cobalamin deficiency.
Journal of the American Animal Hospital Association (2013-03-29)
Sabina Lutz, Adrian C Sewell, Claudia E Reusch, Peter H Kook
ABSTRACT
Juvenile cobalamin deficiency is a rare disease in border collies and its diagnosis requires a high level of clinical suspicion. The goal of this study was to increase awareness of this disease by describing the clinical and laboratory findings in four young border collies with inherited cobalamin deficiency. The median age of the dogs was 11.5 mo (range, 8-42 mo), and two of the four dogs were full siblings. Clinical signs included intermittent lethargy (n = 4), poor body condition (n = 4), odynophagia (n = 2), glossitis (n = 1), and bradyarrhythmia (n = 1). Pertinent laboratory abnormalities were mild to moderate normocytic nonregenerative anemia (n = 3), increased aspartate aminotransferase (AST) activity (n = 3), and mild proteinuria (n = 3). All of the dogs had serum cobalamin levels below the detection limit of the assay, marked methylmalonic aciduria, and hyperhomocysteinemia. Full clinical recovery was achieved in all dogs with regular parenteral cobalamin supplementation, and laboratory abnormalities resolved, except the proteinuria and elevated AST activity persisted. This case series demonstrates the diverse clinical picture of primary cobalamin deficiency in border collies. Young border collies presenting with ambiguous clinical signs should be screened for cobalamin deficiency.
Product Description
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%