Merck
A three-level luminescent response in a pyrene/ferrocene rotaxane.
Organic letters (2012-12-22)
Aurelio Mateo-Alonso, Christian Ehli, Dirk M Guldi, Maurizio Prato
ABSTRACT
A solvent switchable rotaxane equipped with a pyrene stopper and with two ferrocenyl units on the macrocycle is reported, in which three different states, two nondegenerate and one degenerate, can be obtained in different solvents at room temperature. This is accompanied by high contrast changes in fluorescence intensity of the pyrene stopper by the presence of the ferrocenyl moieties on the macrocycle, which quench the emission of pyrene more efficiently with proximity.
Product Description
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Pyrene, 98%
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ferrocene, 98%