Merck
  • Reversible off-on fluorescence probe for hypoxia and imaging of hypoxia-normoxia cycles in live cells.
Reversible off-on fluorescence probe for hypoxia and imaging of hypoxia-normoxia cycles in live cells.
Journal of the American Chemical Society (2012-11-20)
Shodai Takahashi, Wen Piao, Yuriko Matsumura, Toru Komatsu, Tasuku Ueno, Takuya Terai, Toshiaki Kamachi, Masahiro Kohno, Tetsuo Nagano, Kenjiro Hanaoka
ABSTRACT
We report a fully reversible off-on fluorescence probe for hypoxia. The design employs QSY-21 as a Förster resonance energy transfer (FRET) acceptor and cyanine dye Cy5 as a FRET donor, based on our finding that QSY-21 undergoes one-electron bioreduction to the radical under hypoxia, with an absorbance decrease at 660 nm. At that point, FRET can no longer occur, and the dye becomes strongly fluorescent. Upon recovery of normoxia, the radical is immediately reoxidized to QSY-21, with loss of fluorescence due to restoration of FRET. We show that this probe, RHyCy5, can monitor repeated hypoxia-normoxia cycles in live cells.
Product Description
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, certified by the Biological Stain Commission
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy
Sigma-Aldrich
Malachite Green solution, for microscopy