Merck
  • Fundamental characteristics study of anion-exchange PVDF-SiO(2) membranes.
Fundamental characteristics study of anion-exchange PVDF-SiO(2) membranes.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-10-04)
Xingtao Zuo, Wenxin Shi, Shuili Yu, Jiajie He
ABSTRACT
A new type of poly(vinylidene fluoride)(PVDF)-SiO(2) hybrid anion-exchange membrane was prepared by blending method. The anion-exchange groups were introduced by the reaction of epoxy groups with trimethylamine (TMA). Contact angle between water and the membrane surface was measured to characterize the hydrophilicity change of the membrane surface. The effects of nano-sized SiO(2) particles in the membrane-forming materials on the membrane mechanical properties and conductivity were also investigated. The experimental results indicated that PVDF-SiO(2) anion-exchange membranes exhibited better water content, ion-exchange capacity, conductivity and mechanic properties, and so may find potential applications in alkaline membrane fuel cells and water treatment processes.
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~534,000 by GPC, powder
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~180,000 by GPC, average Mn ~71,000, beads or pellets
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~275,000 by GPC, average Mn ~107,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~530,000, pellets