Merck
A dual-targeting hybrid platinum(IV) prodrug for enhancing efficacy.
Chemical communications (Cambridge, England) (2012-09-27)
Haihua Xiao, Lesan Yan, Yu Zhang, Ruogu Qi, Wenliang Li, Rui Wang, Shi Liu, Yubin Huang, Yuxin Li, Xiabin Jing
ABSTRACT
A multifunctional hybrid platinum(IV) prodrug, which consists of both the mitochondria-targeting drug DCA and the DNA-crosslinking drug cisplatin, was synthesized and tethered to a carrier polymer to further self-assemble into micelles for intracellular delivery.
Sigma-Aldrich
DCA Deblock (0.36M dichloroacetic acid in toluene)
Supelco
Dichloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%