Merck
  • Viscerosomatic facilitation in a subset of IBS patients, an effect mediated by N-methyl-D-aspartate receptors.
Viscerosomatic facilitation in a subset of IBS patients, an effect mediated by N-methyl-D-aspartate receptors.
The journal of pain : official journal of the American Pain Society (2012-09-11)
G Nicholas Verne, Donald D Price, Christopher S Callam, Buyi Zhang, Josh Peck, QiQi Zhou
ABSTRACT
Irritable bowel syndrome (IBS) is a common gastrointestinal disorder in which the pathophysiological mechanisms of the pain and hypersensitivity are incompletely understood. IBS patients frequently complain of pain in body regions somatotopically distinct from the gut, suggesting involvement of central hyperalgesic mechanisms. We tested the role of tonic peripheral impulse input by using both repetitive thermal stimuli to the leg and repetitive stimuli to the rectum. Changes in thermal/visceral pain sensitivity after nociceptive thermal/visceral repetitive stimulation were determined. A subset of IBS patients showed enhanced rectal/thermal pain sensitivity after repetitive thermal/rectal stimulation, respectively. IBS patients then received 60 mg dextromethorphan and placebo (diphenhydramine) in a randomized, double-blind, crossover trial. The results showed that 1) a subset of IBS patients had increased visceral/cutaneous hypersensitivity following a series of repetitive nociceptive stimuli and that 2) this increased pain sensitivity was blocked by administration of dextromethorphan. This is the first human study indicating that repetitive stimulation enhances a bidirectional mechanism of secondary hyperalgesia due to viscerosomatic facilitation in IBS patients. These unique findings elucidate mechanisms of somatic hypersensitivity in IBS patients and support an etiologic basis for abnormal N-methyl-D-aspartate receptor mechanisms that may be the target of future therapies for IBS. Repetitive stimulation enhances a bidirectional mechanism of secondary hyperalgesia due to viscerosomatic convergence in IBS patients. The findings elucidate unique mechanisms of somatic/visceral hypersensitivity in a subset of IBS patients and further support an etiologic basis for abnormal N-methyl-D-aspartate receptor mechanisms that may be future targets of therapies for IBS.
Sigma-Aldrich
Dextromethorphan hydrobromide, meets USP testing specifications
Supelco
Dextromethorphan HBr, certified reference material, pharmaceutical secondary standard
Supelco
Dextromethorphan solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dextromethorphan hydrobromide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dextromethorphan hydrobromide monohydrate, ≥99% (TLC)