Merck
  • Cross-complementation study of the flagellar type III export apparatus membrane protein FlhB.
Cross-complementation study of the flagellar type III export apparatus membrane protein FlhB.
PloS one (2012-09-07)
Clive S Barker, Fadel A Samatey
ABSTRACT
The bacterial type III export apparatus is found in the flagellum and in the needle complex of some pathogenic Gram-negative bacteria. In the needle complex its function is to secrete effector proteins for infection into Eukaryotic cells. In the bacterial flagellum it exports specific proteins for the building of the flagellum during its assembly. The export apparatus is composed of about five membrane proteins and three soluble proteins. The mechanism of the export apparatus is not fully understood. The five membrane proteins are well conserved and essential. Here a cross-complementation assay was performed: substituting in the flagellar system of Salmonella one of these membrane proteins, FlhB, by the FlhB ortholog from Aquifex aeolicus (an evolutionary distant hyperthermophilic bacteria) or a chimeric protein (AquSalFlhB) made by the combination of the trans-membrane domain of A. aeolicus FlhB with the cytoplasmic domain of Salmonella FlhB dramatically reduced numbers of flagella and motility. From cells expressing the chimeric AquSalFlhB protein, suppressor mutants with enhanced motility were isolated and the mutations were identified using whole genome sequencing. Gain-of-function mutations were found in the gene encoding FlhA, another membrane protein of the type III export apparatus. Also, mutations were identified in genes encoding 4-hydroxybenzoate octaprenyltransferase, ubiquinone/menaquinone biosynthesis methyltransferase, and 4-hydroxy-3-methylbut-2-en-1-yl diphosphate synthase, which are required for ubiquinone biosynthesis. The mutations were shown by reversed-phase high performance liquid chromatography to reduce the quinone pool of the cytoplasmic membrane. Ubiquinone biosynthesis could be restored for the strain bearing a mutated gene for 4-hydroxybenzoate octaprenyltransferase by the addition of excess exogenous 4-hydroxybenzoate. Restoring the level of ubiquinone reduced flagella biogenesis with the AquSalFlhB chimera demonstrating that the respiratory chain quinone pool is responsible for this phenomenon.
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium 4-hydroxybenzoate, analytical standard