Merck
Assays for cytokines using aptamers.
Methods (San Diego, Calif.) (2006-02-24)
Jeffrey W Guthrie, Camille L A Hamula, Hongquan Zhang, X Chris Le
ABSTRACT
Aptamers are short nucleic acid sequences that are used as ligands to bind their targets with high affinity. They are generated via the combinatorial chemistry procedure systematic evolution of ligands by exponential enrichment (SELEX). Aptamers have shown much promise towards detection of a variety of protein targets, including cytokines. Specifically, for the determination of cytokines and growth factors, several assays making use of aptamers have been developed, including aptamer-based enzyme-linked immunosorbent assays, antibody-linked oligonucleotide assay, fluorescence (anisotropy and resonance energy transfer) assays, and proximity ligation assays. In this article, the concept of aptamer selection using SELEX and the assay formats using aptamers for the detection of cytokines are discussed.
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Pricing and availability is not currently available.