Merck
  • Sorption of Cr(III) ion from aqueous solution by two kinds of modified diatomite.
Sorption of Cr(III) ion from aqueous solution by two kinds of modified diatomite.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-07-26)
Er Li, Xiangying Zeng
ABSTRACT
Raw diatomite modified by microemulsion (DMM) and manganese oxide (MnD) were used for the removal of Cr(III) ions from aqueous solution. The characteristics and performance of these two types of modified diatomite on Cr(III) ion adsorption were compared. The results indicate that the Cr(III) ion adsorption capacities of diatomite were considerably improved after modifications by manganese oxide (MnO) and microemulsion. The surface area of MnD was increased because of the formation of MnO on the diatomite surface, and that of DMM was promoted owing to the existence of the hydrolyzed aromatic acid. Because of the stronger surface ionized function, the adsorption performance of Cr(III) ions in DMM was better than that in MnD. Within the experimental range of pH (i.e. 2.2-6.3), the Cr(III) ion removal of DMM (35-70%) was higher than that of MnD (33-59%) owing to the different electrostatic forces between the Cr(III) ion and the surface of the modified diatomite. The Cr(III) ion removal in MnD and DMM was improved with the increase of synthetic solution concentration in volumes from 0 to 2,500 mL.
Product Description
Supelco
Celite® 545 AW, reagent grade
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Diatomaceous earth