Merck
Pichia fermentans dimorphic changes depend on the nitrogen source.
Fungal biology (2012-07-04)
Maria Lina Sanna, Severino Zara, Giacomo Zara, Quirico Migheli, Marilena Budroni, Ilaria Mannazzu
ABSTRACT
Pichia fermentans DiSAABA 726 is a biofilm-forming yeast that undergoes dimorphic transition. Under yeast-like morphology it controls brown rot caused by Monilia spp. on apple fruit, while under pseudohyphal form, it shows pathogenic behaviour itself on peach fruit. The present study investigates the nutritional factors that induce and separate yeast-like and pseudohyphal morphologies under laboratory conditions. We show that P. fermentans DiSAABA 726 produces mainly yeast-like cells on media containing millimolar concentrations of urea and diammonium phosphate, and forms pseudohyphae at micromolar concentrations of these two salts. With ammonium sulphate, yeast-like or pseudohyphal morphology depends on the N concentration and the pH of the culture media. Amino acids such as methionine, valine, and phenylalanine invariably induce pseudohyphal morphology irrespective of the N concentration and the pH of the culture media. Methionol, 1-butanol, isobutanol, and isopropanol induce pseudohyphal growth, while phenylethanol and isoamyl alcohol fail to induce the formation of filaments. Thus, the morphogenesis of P. fermentans DiSAABA 726 depends more on the nitrogen source than on the N concentration, and is regulated by the quorum-sensing molecules that are generally produced from amino-acid assimilation under nitrogen starvation.
Product Description
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
Ammonium phosphate monobasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%