Merck
  • Extremely thin bilayer electrolyte for solid oxide fuel cells (SOFCs) fabricated by chemical solution deposition (CSD).
Extremely thin bilayer electrolyte for solid oxide fuel cells (SOFCs) fabricated by chemical solution deposition (CSD).
Advanced materials (Deerfield Beach, Fla.) (2012-06-01)
Eun-Ok Oh, Chin-Myung Whang, Yu-Ri Lee, Sun-Young Park, Dasari Hari Prasad, Kyung Joong Yoon, Ji-Won Son, Jong-Ho Lee, Hae-Weon Lee
ABSTRACT
An extremely thin bilayer electrolyte consisting of yttria-stabilized zirconia (YSZ) and gadolinia-doped ceria (GDC) is successfully fabricated on a sintered NiO-YSZ substrate. Major processing flaws are effectively eliminated by applying local constraints to YSZ nanoparticles, and excellent open circuit voltage and cell performance are demonstrated in a solid oxide fuel cell (SOFC) at intermediate operating temperatures.
Sigma-Aldrich
Yttrium(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, nanopowder, <50 nm particle size
Sigma-Aldrich
Yttrium(III) oxide, dispersion, 10 wt. % in isopropanol, nanoparticles, <100 nm (DLS), ≥99.9% trace metals basis