Merck
  • High-temperature rate constant determination for the reaction of OH with iso-butanol.
High-temperature rate constant determination for the reaction of OH with iso-butanol.
The journal of physical chemistry. A (2012-04-21)
Genny A Pang, Ronald K Hanson, David M Golden, Craig T Bowman
ABSTRACT
This work presents the first direct experimental study of the rate constant for the reaction of OH with iso-butanol (2-methyl-1-propanol) at temperatures from 907 to 1147 K at near-atmospheric pressures. OH time-histories were measured behind reflected shock waves using a narrow-linewidth laser absorption method during reactions of dilute mixtures of tert-butylhydroperoxide (as a fast source of OH) with iso-butanol in excess. The title reaction's overall rate constant (OH + iso-butanol →(k(overall)) all products) minus the rate constant for the β-radical-producing channel (OH + iso-butanol →(k(β)) 1-hydroxy-2-methyl-prop-2-yl radical + H(2)O) was determined from the pseudo-first-order rate of OH decay. A two-parameter Arrhenius fit of the experimentally determined rate constant in the current temperature range yields the expression (k(overall) - k(β)) = 1.84 × 10(-10) exp(-2350/T[K]) cm(3) molecule(-1) s(-1). A recommendation for the overall rate constant, including k(β), is made, and comparisons of the results to rate constant recommendations from the literature are discussed.
Product Description
Supelco
2-Methyl-1-propanol, analytical standard
Sigma-Aldrich
2-Methyl-1-propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Methyl-1-propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Methyl-1-propanol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
2-Methyl-1-propanol, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Isobutyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Isobutyl alcohol, ≥99%, FCC, FG