Merck
  • Removal of zearalenone toxin from synthetics gastric and body fluids using talc and diatomite: a batch kinetic study.
Removal of zearalenone toxin from synthetics gastric and body fluids using talc and diatomite: a batch kinetic study.
Colloids and surfaces. B, Biointerfaces (2012-02-14)
Myroslav Sprynskyy, Renata Gadzała-Kopciuch, Karolina Nowak, Bogusław Buszewski
ABSTRACT
Adsorption kinetics of zearalenone (ZEA) toxin from synthetic gastric fluid (SGF) and synthetic body fluid (SBF) by talc and diatomite was studied in the batch experiments. Chemical composition, morphology and structure of the used adsorbents were examined by scanning electron microscopy, FTIR spectroscopy and low-temperature nitrogen adsorption/desorption method. High performance liquid chromatography (HPLC) method was used for ZEA determining. The study results showed that ZEA is more effectively adsorbed on the talc (73% and 54% from SGF and SBF respectively). The efficiency on the diatomite was lower (53% and 42% from SGF and SBF respectively). The first order kinetics model was applied to describe the adsorption process. Rate of the ZEA adsorption from SGF is very rapid initially with about 95% of amount of the toxin adsorbed during first 5 min, while ZEA is adsorbed from SBF in two steps. The values of determined Gibbs free energy of adsorption (from -13 to -17 kJ/mol) indicated that adsorption of ZEA toxin by the both adsorbents are spontaneous and exothermic.
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Diatomaceous earth
Supelco
Celite® 545 AW, reagent grade
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications