Merck
  • SnCl2-catalyzed selective atom economic imino Diels-Alder reaction: synthesis of 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)quinolines.
SnCl2-catalyzed selective atom economic imino Diels-Alder reaction: synthesis of 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)quinolines.
The Journal of organic chemistry (2012-01-11)
Rajendran Suresh, Shanmugam Muthusubramanian, Rajendran Senthilkumaran, Govindaswamy Manickam
ABSTRACT
The synthesis of 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)quinolines by a SnCl(2)-catalyzed multicomponent reaction has been described. The reaction proceeds chemo- and regioselectively in an atom-economic way, generating a library of 24 quinoline derivatives.
Sigma-Aldrich
Tin(II) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
Supelco
Tin(II) chloride dihydrate, suitable for AAS
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.97% trace metals basis
Sigma-Aldrich
Tin(II) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride dihydrate, reagent grade, 98%
Sigma-Aldrich
Tin(II) chloride dihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Tin(II) chloride dihydrate, ACS reagent, 98%
Sigma-Aldrich
Tin(II) chloride, reagent grade, 98%