Merck
  • Synthesis of arylnaphthalene lignan scaffold by gold-catalyzed intramolecular sequential electrophilic addition and benzannulation.
Synthesis of arylnaphthalene lignan scaffold by gold-catalyzed intramolecular sequential electrophilic addition and benzannulation.
The Journal of organic chemistry (2011-10-28)
Vanajakshi Gudla, Rengarajan Balamurugan
ABSTRACT
An intramolecular approach to generate compounds containing an arylnaphthalene lignan scaffold in high yields is presented. It involves a sequential intramolecular electrophilic attack of carbonyl on arylalkyne followed by benzannulation catalyzed by gold salt. AuCl(3) in combination with AgSbF(6) works better to effect this transformation. Selected products have been converted into arylnaphthalene lactone natural products such as justicidin E, taiwanin C, and retrojusticidin B.
Sigma-Aldrich
Gold(I) chloride, 99.9% trace metals basis
Sigma-Aldrich
Gold(III) chloride, 99%
Sigma-Aldrich
Gold(III) chloride, ≥99.99% trace metals basis