Merck
  • Primary amine/(+)-CSA salt-promoted organocatalytic conjugate addition of nitro esters to enones.
Primary amine/(+)-CSA salt-promoted organocatalytic conjugate addition of nitro esters to enones.
Organic letters (2010-04-24)
Chen Liu, Yixin Lu
ABSTRACT
The combination of 9-amino-9-deoxy-epi-cinchonine and (+)-CSA resulted in a novel primary amine-based organocatalyst for effective iminium activation of alpha,beta-unsaturated ketones. Such a catalytic system could catalyze the conjugate addition of nitroacetate to enones in a highly enantioselective manner, affording the desired adducts in high yields and with up to 99% ee.
Sigma-Aldrich
(+)-Camphor-10-sulfonic acid, purum, ≥98.0% (T)
Sigma-Aldrich
(1S)-(+)-10-Camphorsulfonic acid, 99%
Sigma-Aldrich
(1R)-(−)-10-Camphorsulfonic acid, 98%
Sigma-Aldrich
(−)-Camphor-10-sulfonic acid, purum, ≥98.0% (T)
Sigma-Aldrich
Camphor-10-sulfonic acid (β), 98%