Merck
Preparation and properties of lithium and sodium peroxynitrite.
Chemical research in toxicology (2009-06-24)
Manuel Sturzbecher-Höhne, Reinhard Kissner, Thomas Nauser, Willem H Koppenol
ABSTRACT
We describe the preparation of aqueous solutions of LiONOO and NaONOO from (Me4N)ONOO. An aqueous solution of analytically pure, commercially available (Me4N)ONOO is applied to an Amberlyst 15 column at 4 degrees C, and the Me4N+ is rapidly (in 20 min) exchanged against Li+ or Na+. The exchanged peroxynitrite is produced in yields of ca. 90% with a nitrite content of 5-6%.
Sigma-Aldrich
Lithium hydroxide ChemBeads
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Sigma-Aldrich
Lithium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Lithium hydroxide monohydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Lithium hydroxide monohydrate, BioXtra, 98.5-101.5% (titration)
Sigma-Aldrich
Lithium hydroxide monohydrate, ACS reagent, ≥98.0%