Merck
  • Synthesis of hexagonal nickel hydroxide nanosheets by exfoliation of layered nickel hydroxide intercalated with dodecyl sulfate ions.
Synthesis of hexagonal nickel hydroxide nanosheets by exfoliation of layered nickel hydroxide intercalated with dodecyl sulfate ions.
Journal of the American Chemical Society (2008-10-07)
Shintaro Ida, Daisuke Shiga, Michio Koinuma, Yasumichi Matsumoto
ABSTRACT
One-nanometer-thick nickel hydroxide nanosheets were prepared by exfoliation of layered nickel hydroxides intercalated with dodecyl sulfate (DS) ions. The shape of the nanosheets was hexagonal, as was that of the layered nickel hydroxides intercalated with DS ions. The nickel hydroxide nanosheets exhibited charge-discharge properties in strong alkaline electrolyte. The morphology of the nanosheet changed during the electrochemical reaction.
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Ammonium lauryl sulfate solution, ~30% in H2O (T)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Nickel(II) hydroxide
Sigma-Aldrich
Ammonium lauryl sulfate solution, ~30% in H2O