Merck
Carbonate assignment and calibration in the Raman spectrum of apatite.
Calcified tissue international (2007-06-07)
Ayorinde Awonusi, Michael D Morris, Mary M J Tecklenburg
ABSTRACT
A series of apatites with varying carbonate levels was prepared in order to assign the carbonate bands and calibrate for Raman analysis of natural materials. Overlap of carbonate bands with phosphate peaks was resolved by curve fitting. A peak at 1,071 cm(-1) was assigned to a combination of the carbonate nu(1) mode at 1,070 cm(-1) with a phosphate nu(3) mode at 1,076 cm(-1). In addition, the carbonate nu(4) mode was identified in apatite samples with >4% carbonate. The carbonate nu(4) bands at 715 and 689 cm(-1) identify the samples as B-type carbonated apatite. The carbonate content of apatite was calibrated to a carbonate Raman band, and the method was used to determine the carbonate content of a sample of bovine cortical bone, 7.7 +/- 0.4%.
Sigma-Aldrich
Lithium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Lithium hydroxide monohydrate, BioXtra, 98.5-101.5% (titration)
Sigma-Aldrich
Lithium hydroxide monohydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Lithium hydroxide monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%