Merck
  • Pediatric susceptibility to 18 industrial chemicals: a comparative analysis of newborn with young animals.
Pediatric susceptibility to 18 industrial chemicals: a comparative analysis of newborn with young animals.
Regulatory toxicology and pharmacology : RTP (2006-12-13)
R Hasegawa, M Hirata-Koizumi, M Dourson, A Parker, A Hirose, S Nakai, E Kamata, M Ema
ABSTRACT
We comprehensively re-analyzed the toxicity data for 18 industrial chemicals from repeated oral exposures in newborn and young rats, which were previously published. Two new toxicity endpoints specific to this comparative analysis were identified, the first, the presumed no observed adverse effect level (pNOAEL) was estimated based on results of both main and dose-finding studies, and the second, the presumed unequivocally toxic level (pUETL) was defined as a clear toxic dose giving similar severity in both newborn and young rats. Based on the analyses of both pNOAEL and pUETL ratios between the different ages, newborn rats demonstrated greater susceptibility (at most 8-fold) to nearly two thirds of these 18 chemicals (mostly phenolic substances), and less or nearly equal sensitivity to the other chemicals. Exceptionally one chemical only showed toxicity in newborn rats. In addition, Benchmark Dose Lower Bound (BMDL) estimates were calculated as an alternative endpoint. Most BMDLs were comparable to their corresponding pNOAELs and the overall correlation coefficient was 0.904. We discussed how our results can be incorporated into chemical risk assessment approaches to protect pediatric health from direct oral exposure to chemicals.
Sigma-Aldrich
Trityl chloride, 97%
Sigma-Aldrich
Trityl chloride, purum, ≥97.0% (HPLC), ≥97.0% (AT)