Merck
  • Application of a new electrolyte circulation method for the ex situ electrokinetic bioremediation of a laboratory-prepared pentadecane contaminated kaolinite.
Application of a new electrolyte circulation method for the ex situ electrokinetic bioremediation of a laboratory-prepared pentadecane contaminated kaolinite.
Journal of hazardous materials (2005-02-22)
Sang-Joon Kim, Ji-Yeon Park, You-Jin Lee, Jae-Young Lee, Ji-Won Yang
ABSTRACT
Ex situ electrokinetic (EK) bioremediation of a laboratory-prepared pentadecane-contaminated kaolinite was carried out. Extraneous bacteria and ionic nutrients were continuously supplied to the soil specimen by a new electrolyte circulation method, which controlled electrical pH change of electrolyte solution to keep bacterial activity. During the EK bioremediation the anode region showed the highest colony forming unit (CFU) due to electrical attraction between anode and bacteria. Simultaneous increases of CFU and uniform pentadecane removal in most soil regions demonstrated that electro-osmosis as well as electrophoresis affected the bacterial transport in soil. At 3.13 mA/cm2, increase in soil temperature to above 45 degrees C inhibited bacterial activity, which caused the decrease of removal efficiency. The removal amount of pentadecane increased with initial pentadecane concentration at the same current densities (0.63 and 1.88 mA/cm2) because of the increased amount of weakly bound pentadecane onto the soil surface. The highest removal efficiency (77.6%) was obtained at 0.63 mA/cm2 for 1000 mg/kg pentadecane after 14 days. Consequently, the present methods of EK bioremediation demonstrated superiority over the conventional bioremediation, which had inherent demerits of slow degradation and low removal efficiency.
Sigma-Aldrich
Pentadecane, ≥98.0% (GC)
Sigma-Aldrich
Pentadecane, ≥99%
Supelco
Pentadecane, analytical standard