Merck
  • Synaptic-like transmission between neural axons and arteriolar smooth muscle cells drives cerebral neurovascular coupling.
Synaptic-like transmission between neural axons and arteriolar smooth muscle cells drives cerebral neurovascular coupling.
Nature neuroscience (2024-01-04)
Dongdong Zhang, Jiayu Ruan, Shiyu Peng, Jinze Li, Xu Hu, Yiyi Zhang, Tianrui Zhang, Yaping Ge, Zhu Zhu, Xian Xiao, Yunxu Zhu, Xuzhao Li, Tingbo Li, Lili Zhou, Qingzhu Gao, Guoxiao Zheng, Bingrui Zhao, Xiangqing Li, Yanming Zhu, Jinsong Wu, Wensheng Li, Jingwei Zhao, Woo-Ping Ge, Tian Xu, Jie-Min Jia
ABSTRACT
Neurovascular coupling (NVC) is important for brain function and its dysfunction underlies many neuropathologies. Although cell-type specificity has been implicated in NVC, how active neural information is conveyed to the targeted arterioles in the brain remains poorly understood. Here, using two-photon focal optogenetics in the mouse cerebral cortex, we demonstrate that single glutamatergic axons dilate their innervating arterioles via synaptic-like transmission between neural-arteriolar smooth muscle cell junctions (NsMJs). The presynaptic parental-daughter bouton makes dual innervations on postsynaptic dendrites and on arteriolar smooth muscle cells (aSMCs), which express many types of neuromediator receptors, including a low level of glutamate NMDA receptor subunit 1 (Grin1). Disruption of NsMJ transmission by aSMC-specific knockout of GluN1 diminished optogenetic and whisker stimulation-caused functional hyperemia. Notably, the absence of GluN1 subunit in aSMCs reduced brain atrophy following cerebral ischemia by preventing Ca2+ overload in aSMCs during arteriolar constriction caused by the ischemia-induced spreading depolarization. Our findings reveal that NsMJ transmission drives NVC and open up a new avenue for studying stroke.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-NeuN purified Antibody, from guinea pig, purified by affinity chromatography
Sigma-Aldrich
Anti-Tau-1 Antibody, clone PC1C6, Alexa Fluor 488 Conjugate, clone PC1C6, from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 7E3-1B8, clone 7E3-1B8, Chemicon®, from mouse