Merck
  • Classification of Cryptosporidium species from patients with sporadic cryptosporidiosis by use of sequence-based multilocus analysis following mutation scanning.
Classification of Cryptosporidium species from patients with sporadic cryptosporidiosis by use of sequence-based multilocus analysis following mutation scanning.
Journal of clinical microbiology (2008-05-02)
Aaron R Jex, Aradhana Pangasa, Bronwyn E Campbell, Margaret Whipp, Geoff Hogg, Martha I Sinclair, Melita Stevens, Robin B Gasser
ABSTRACT
In the present study, we analyzed genetic variation in Cryptosporidium species from humans (n = 62) with clinical cryptosporidiosis in South Australia. Sequence variation was assessed in regions within the small subunit of nuclear rRNA (p-SSU), the 70-kDa heat shock protein (p-hsp70), and the 60-kDa glycoprotein (p-gp60) genes by employing single-strand conformation polymorphism analysis and sequencing. Based on the analyses of p-SSU and p-hsp70, Cryptosporidium hominis (n = 38) and Cryptosporidium parvum (n = 24) were identified. The analysis of p-gp60 revealed eight distinct subgenotypes, classified as C. hominis IaA17R1 (n = 3), IbA9G3R2 (n = 14), IbA10G2R2 (n = 20), and IfA12G1R1 (n = 1), as well as C. parvum IIaA18G3R1 (n = 15), IIaA20G3R1 (n = 6), IIaA22G4R1 (n = 2), and IIcA5G3R2 (n = 1). Subgenotypes IaA17R1 and IIaA22G4R1 are new. Of the six other subgenotypes, IbA10G2R2, IIaA18G3R1, IIaA20G3R1, and IIcA5G3R2 were reported previously from the state of Victoria. This is the fourth record in Australia of C. parvum subgenotype IIaA18G3R1 from humans, which, to date, has been isolated only from cattle in other countries. This subgenotype might be a significant contributor to sporadic human cryptosporidiosis and may indicate a greater zoonotic contribution to the infection of humans in the area of study. Comparative analyses revealed, for the first time, the differences in the genetic makeup of Cryptosporidium populations between two relatively close, major metropolitan cities.
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000