Merck
  • Polyphenolic compounds with biological activity in guabiroba fruits (Campomanesia xanthocarpa Berg.) by comprehensive two-dimensional liquid chromatography.
Polyphenolic compounds with biological activity in guabiroba fruits (Campomanesia xanthocarpa Berg.) by comprehensive two-dimensional liquid chromatography.
Electrophoresis (2020-08-12)
Stefany Grutzmann Arcari, Katia Arena, Jeferson Kolling, Paloma Rocha, Paola Dugo, Luigi Mondello, Francesco Cacciola
ABSTRACT
This study aimed to evaluate the polyphenolic composition along with the biological activity of guabiroba (Campomanesia xanthocarpa Berg.) fruits using comprehensive two-dimensional liquid chromatography (LC × LC). A simplex centroid design comprising three solvents (methanol, 2% acetic acid, and acetonitrile) was used to optimize the extraction mixture for polyphenols from ripe and unripe guabiroba fruits. A quantitative LC × LC platform was proposed to characterize the guabiroba extracts using a RP-Amide column and a C18 column in the first and second dimensions, respectively. Antidiabetic properties, using in vitro enzyme assay models and in vivo antioxidant activity with the eukaryote model Saccharomyces cerevisiae, was measured. Total phenolics compounds were more efficiently extracted with 2% acetic acid solution and acetonitrile (50:50, v/v). A total of 37 different compounds were identified and quantified using the proposed LC × LC method (linearity ranging from 0.9990 to 0.9994, intra- and interday precision from 0.40 to 10.57% and, accuracy from 81.89 to 108.98%). Significant differences were observed between ripe and unripe guabiroba fruits, especially for the compounds geraldone and methyl galangin isomer. Guabiroba fruits showed significant antidiabetic and antioxidant properties and may be potentially adopted as part of dietary strategies in the management of early stages of type 2 diabetes and associated complications.
Product Description
Supelco
Quercetin 3-glucoside, analytical standard
Supelco
trans-Sinapic acid, analytical standard
Supelco
(+)-Catechin, analytical standard
Supelco
Hesperidin, analytical standard
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
(±)-Naringenin, analytical standard
Supelco
Apigenin, analytical standard
Supelco
Kaempferol, analytical standard
Supelco
(−)-Epicatechin, analytical standard