Merck
  • Amplified stimulated emission in upconversion nanoparticles for super-resolution nanoscopy.
Amplified stimulated emission in upconversion nanoparticles for super-resolution nanoscopy.
Nature (2017-02-23)
Yujia Liu, Yiqing Lu, Xusan Yang, Xianlin Zheng, Shihui Wen, Fan Wang, Xavier Vidal, Jiangbo Zhao, Deming Liu, Zhiguang Zhou, Chenshuo Ma, Jiajia Zhou, James A Piper, Peng Xi, Dayong Jin
ABSTRACT
Lanthanide-doped glasses and crystals are attractive for laser applications because the metastable energy levels of the trivalent lanthanide ions facilitate the establishment of population inversion and amplified stimulated emission at relatively low pump power. At the nanometre scale, lanthanide-doped upconversion nanoparticles (UCNPs) can now be made with precisely controlled phase, dimension and doping level. When excited in the near-infrared, these UCNPs emit stable, bright visible luminescence at a variety of selectable wavelengths, with single-nanoparticle sensitivity, which makes them suitable for advanced luminescence microscopy applications. Here we show that UCNPs doped with high concentrations of thulium ions (Tm3+), excited at a wavelength of 980 nanometres, can readily establish a population inversion on their intermediate metastable 3H4 level: the reduced inter-emitter distance at high Tm3+ doping concentration leads to intense cross-relaxation, inducing a photon-avalanche-like effect that rapidly populates the metastable 3H4 level, resulting in population inversion relative to the 3H6 ground level within a single nanoparticle. As a result, illumination by a laser at 808 nanometres, matching the upconversion band of the 3H4 → 3H6 transition, can trigger amplified stimulated emission to discharge the 3H4 intermediate level, so that the upconversion pathway to generate blue luminescence can be optically inhibited. We harness these properties to realize low-power super-resolution stimulated emission depletion (STED) microscopy and achieve nanometre-scale optical resolution (nanoscopy), imaging single UCNPs; the resolution is 28 nanometres, that is, 1/36th of the wavelength. These engineered nanocrystals offer saturation intensity two orders of magnitude lower than those of fluorescent probes currently employed in stimulated emission depletion microscopy, suggesting a new way of alleviating the square-root law that typically limits the resolution that can be practically achieved by such techniques.
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4,Yb,Nd, PEG-COOH modified core-shell, fluorescence λex 808 nm, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4-Yb,Er@NaYF4, fluorescence λex 980 nm, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4 Yb, Nd, PEG-NH2 modified core-shell, fluorescence λex 808 nm, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4-Yb,Tm@NaYF4, 980 excitation, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, PEG-NH2 modified core-shell, 808 excitation, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4:Yb,Er@NaYF4, Oil soluble core-shell, fluorescence λex 980, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4:Yb,Tm@NaYF4, Oil soluble core shell, 980 excitation, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, Oil soluble core shell,808 excitation, green
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Er@NaYF4, PEG-NH2 modified core-shell, 980 excitation, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4,Yb,Nd, Oil soluble core shell upconversion nanoparticle, 808 excited, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Er@NaYF4, PEG-COOH modified core-shell, fluorescence λex 980 nm, green light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4,Yb,Tm@NaYF4,Yb,Nd, 808 excited, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4,Yb,Er@NaYF4,Yb,Nd, 808 excited, green
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, PEG-COOH modified core-shell, 808 excitation, green light