Merck
Catalase and its mysteries.
Progress in biophysics and molecular biology (2018-03-14)
Hessam Sepasi Tehrani, Ali Akbar Moosavi-Movahedi
ABSTRACT
Catalase is one of the firsts in every realm of biological sciences. At the same time it also has a number of unusual features. It has one of the highest turnover numbers of all enzymes. It is essential for neutralizing the noxious hydrogen peroxide both in the nature and the various industries such as dairy, textile and pharmaceutics. It also has the merit of being one of the first protein crystals to be isolated. Ironically its three-dimensional structure was discerned some forty years later. However through the times this senile enzyme has continued to intrigue the scientists by surprising facts and phenomena, such as peculiar interweaving of subunits and remarkable thermal stability. It is also known for suicide inactivation by its own substrate. Catalase is known to be implicated in various medical scenarios and its levels have served as a marker in that capacity. It has even been incorporated into several pharmaceuticals. This review strives to clarify these perspectives. It also draws attention to the biophysical contributions offered by thermodynamics and kinetics in these discoveries. The ultimate aim of this review, however, is to state that the venerable catalase will continue to bewilder us with its mysteries well into the twenty-first century.
Sigma-Aldrich
Catalase from Micrococcus lysodeikticus, solution, dark brown
Pricing and availability is not currently available.