Merck
  • A switch in pdgfrb+ cell-derived ECM composition prevents inhibitory scarring and promotes axon regeneration in the zebrafish spinal cord.
A switch in pdgfrb+ cell-derived ECM composition prevents inhibitory scarring and promotes axon regeneration in the zebrafish spinal cord.
Developmental cell (2021-01-08)
Vasiliki Tsata, Stephanie Möllmert, Christine Schweitzer, Julia Kolb, Conrad Möckel, Benjamin Böhm, Gonzalo Rosso, Christian Lange, Mathias Lesche, Juliane Hammer, Gokul Kesavan, Dimitris Beis, Jochen Guck, Michael Brand, Daniel Wehner
ABSTRACT
In mammals, perivascular cell-derived scarring after spinal cord injury impedes axonal regrowth. In contrast, the extracellular matrix (ECM) in the spinal lesion site of zebrafish is permissive and required for axon regeneration. However, the cellular mechanisms underlying this interspecies difference have not been investigated. Here, we show that an injury to the zebrafish spinal cord triggers recruitment of pdgfrb+ myoseptal and perivascular cells in a PDGFR signaling-dependent manner. Interference with pdgfrb+ cell recruitment or depletion of pdgfrb+ cells inhibits axonal regrowth and recovery of locomotor function. Transcriptional profiling and functional experiments reveal that pdgfrb+ cells upregulate expression of axon growth-promoting ECM genes (cthrc1a and col12a1a/b) and concomitantly reduce synthesis of matrix molecules that are detrimental to regeneration (lum and mfap2). Our data demonstrate that a switch in ECM composition is critical for axon regeneration after spinal cord injury and identify the cellular source and components of the growth-promoting lesion ECM.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
PDGFR Tyrosine Kinase Inhibitor IV
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Protease from Streptomyces griseus, powder, BioReagent, suitable for mouse embryo cell culture, ≥3.5 units/mg solid
Sigma-Aldrich
PDGFR Tyrosine Kinase Inhibitor V, The PDGFR Tyrosine Kinase Inhibitor V, also referenced under CAS 347155-76-4, controls the biological activity of PDGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep