Merck
  • Binding of ubiquitin conjugates to proteasomes as visualized with native gels.
Binding of ubiquitin conjugates to proteasomes as visualized with native gels.
Methods in molecular biology (Clifton, N.J.) (2012-02-22)
Suzanne Elsasser, Yuan Shi, Daniel Finley
ABSTRACT
The proteasome is an ATP-dependent molecular machine that degrades proteins through the concerted activity of dozens of subunits. It is the yin to the ribosome's yang, and together these entities mold the protein landscape of the cell. Native gels are generally superior to conventional and affinity purifications for the analytical resolution proteasomal variants, and have thus become a staple of proteasome work. Here, we describe the technique of using native gels to observe proteasomes in complex with ubiquitin conjugates. We discuss the consequences of ubiquitin conjugate length and concentration on the migration of these complexes, the use of this mobility shift to evaluate the relative affinity of mutant proteasomes for ubiquitin conjugates, and the effects of deubiquitinating enzymes and competing ubiquitin-binding proteins on the interactions of ubiquitin conjugates with the proteasome.
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Leupeptin hydrochloride, microbial, ≥70% (HPLC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%