Merck
  • PURPL represses autophagic cell death to promote cutaneous melanoma by modulating ULK1 phosphorylation.
PURPL represses autophagic cell death to promote cutaneous melanoma by modulating ULK1 phosphorylation.
Cell death & disease (2021-11-12)
Shuo Han, Xue Li, Ke Wang, Dingheng Zhu, Bingyao Meng, Jieyu Liu, Xiaoting Liang, Yi Jin, Xingyuan Liu, Qian Wen, Liang Zhou
ABSTRACT
Uncontrolled overactivation of autophagy may lead to autophagic cell death, suppression of which is a pro-survival strategy for tumors. However, mechanisms involving key regulators in modulating autophagic cell death remain poorly defined. Here, we report a novel long noncoding RNA, p53 upregulated regulator of p53 levels (PURPL), functions as an oncogene to promote cell proliferation, colony formation, migration, invasiveness, and inhibits cell death in melanoma cells. Mechanistic studies showed that PURPL promoted mTOR-mediated ULK1 phosphorylation at Ser757 by physical interacting with mTOR and ULK1 to constrain autophagic response to avoid cell death. Loss of PURPL led to AMPK-mediated phosphorylation of ULK1 at Ser555 and Ser317 to over-activate autophagy and induce autophagic cell death. Our results identify PURPL as a key regulator to modulate the activity of autophagy initiation factor ULK1 to repress autophagic cell death in melanoma and may represent a potential intervention target for melanoma therapy.
Sigma-Aldrich
Trypsin Profile IGD Kit
Pricing and availability is not currently available.