Merck
  • Kinetic and docking studies of phenol-based inhibitors of carbonic anhydrase isoforms I, II, IX and XII evidence a new binding mode within the enzyme active site.
Kinetic and docking studies of phenol-based inhibitors of carbonic anhydrase isoforms I, II, IX and XII evidence a new binding mode within the enzyme active site.
Bioorganic & medicinal chemistry (2011-02-02)
Serdar Durdagi, Murat Şentürk, Deniz Ekinci, Halis Türker Balaydın, Süleyman Göksu, Ö İrfan Küfrevioğlu, Alessio Innocenti, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
Carbonic anhydrases (CAs, EC 4.2.1.1) are inhibited by sulfonamides, inorganic anions, phenols, coumarins (acting as prodrugs) and polyamines. A novel class of CA inhibitors (CAIs), interacting with the CA isozymes I, II (cytosolic) and IX, XII (transmembrane, tumor-associated) in a different manner, is reported here. Kinetic measurements allowed us to identify hydroxy-/methoxy-substituted benzoic acids as well as di-/tri-methoxy benzenes as submicromolar-low micromolar inhibitors of the four CA isozymes. Molecular docking studies of a set of such inhibitors within CA I and II allowed us to understand the inhibition mechanism. This new class of inhibitors binds differently compared to all other classes of inhibitors known to date: they were found between the phenol-binding site and the coumarin-binding site, filling thus the middle of the enzyme cavity. They exploit different interactions with amino acid residues and water molecules from the CA active site compared to other classes of inhibitors, offering the possibility to design CAIs with an interesting inhibition profile compared to the clinically used sulfonamides/sulfamates.
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Spermine, BioReagent, suitable for cell culture
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
1,2-Dimethoxybenzene, ≥99%, FG
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
1,2-Dimethoxybenzene, ReagentPlus®, 99%
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
3,4-Dimethoxybenzoic acid, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
1,4-Dimethoxybenzene, 99%, FG
Sigma-Aldrich
1,4-Dimethoxybenzene, ReagentPlus®, 99%
Sigma-Aldrich
3,5-Dihydroxybenzoic acid, 97%