Merck
  • Propofol exerts anticancer activity on hepatocellular carcinoma cells by raising lncRNA DGCR5.
Propofol exerts anticancer activity on hepatocellular carcinoma cells by raising lncRNA DGCR5.
Journal of cellular physiology (2019-09-21)
Yan Sun, Hai Sun
ABSTRACT
Hepatocellular carcinoma is one of the most fatal cancers worldwide. Propofol is an intravenous anesthetic extensively used in clinical. Herein, we tested the anticancer activity of propofol on hepatocellular carcinoma, along with the internal molecular mechanism related to lncRNA DiGeorge syndrome critical region gene 5 (DGCR5). Followed by propofol stimulation, hepatocellular carcinoma Huh-7 and HepG2 cell viability, proliferation, migration, invasion, and apoptosis were tested, respectively. Then, DGCR5 expression levels in hepatocellular carcinoma tissues and cells were measured. sh-DGCR5 was transfected to silence DGCR5 expression. Subsequently, the influence of DGCR5 silence on propofol caused Huh-7 and HepG2 cell viability loss, proliferation inhibition, migration and invasion suppression, apoptosis induction, as well as Raf1/ERK1/2 and Wnt/β-catenin pathways inactivation were assessed, respectively. We discovered that propofol declined Huh-7 and HepG2 cell viability, proliferation, migration and invasion, but increased cell apoptosis. DGCR5 had a relatively lower expression level in hepatocellular carcinoma tissues and cells. Propofol elevated DGCR5 expression in Huh-7 and HepG2 cells. Increased expression of DGCR5 was connected with the anticancer activity of propofol on Huh-7 and HepG2 cells. Besides, propofol repressed Raf1/ERK1/2 and Wnt/β-catenin pathways through elevating DGCR5 expression. In conclusion, the anticancer activity of propofol on hepatocellular carcinoma was verified in this study. Propofol repressed hepatocellular carcinoma Huh-7 and HepG2 cell growth and metastasis at least by elevating DGCR5 and hereafter inactivating Raf1/ERK1/2 and Wnt/β-catenin pathways.
Sigma-Aldrich
StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture
Propofol, European Pharmacopoeia (EP) Reference Standard
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture