Merck
  • Confirmation of five nitrofuran metabolites including nifursol metabolite in meat and aquaculture products by liquid chromatography-tandem mass spectrometry: Validation according to European Union Decision 2002/657/EC.
Confirmation of five nitrofuran metabolites including nifursol metabolite in meat and aquaculture products by liquid chromatography-tandem mass spectrometry: Validation according to European Union Decision 2002/657/EC.
Food chemistry (2020-12-04)
Pierre Guichard, Michel Laurentie, Dominique Hurtaud-Pessel, Eric Verdon
ABSTRACT
LC-MS/MS method for confirmation of nitrofuran metabolites in meat and aquaculture products, including the nifursol metabolite (DNSH), was developed. The nitrofuran metabolites investigated were as follows: 3-amino-2-oxazolidinone (AOZ), 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ), 1-aminohydantoine (AHD), semicarbazide (SEM) and 3,5-dinitrosalicylic acid hydrazide (DNSH). The sample preparation includes a washing step, allowing to analyze only the fraction of protein-bound residues. The final optimized recovery solvent for injection was found to be a mixture of ammonium acetate 2 mM/acetonitrile (60/40 ; v/v). Matrix effects and stability in biological matrix and standard solution for DNSH have been also carried out. Method performances were assessed using criteria of the Decision (EC) No 2002/657 and considering the proposed-for-adoption reference point for action (RPA) of 0.50 µg.kg-1 under Reg 1871/2019. Trueness ranged 86.5%-103.7% and precision ranged 2.0%-6.5% on intra-laboratory reproducibility conditions. Decision limit (CCα) ranged 0.028-0.182 μg.kg-1 and capability of detection limit (CCβ) ranged 0.032-0.233 µg.kg-1.
Sigma-Aldrich
2-Nitrofuran, 97%
Supelco
AOZ, VETRANAL®, analytical standard