Merck
  • Repurposing the Antidepressant Sertraline as SHMT Inhibitor to Suppress Serine/Glycine Synthesis-Addicted Breast Tumor Growth.
Repurposing the Antidepressant Sertraline as SHMT Inhibitor to Suppress Serine/Glycine Synthesis-Addicted Breast Tumor Growth.
Molecular cancer therapeutics (2020-11-19)
Shauni Lien Geeraerts, Kim Rosalie Kampen, Gianmarco Rinaldi, Purvi Gupta, Mélanie Planque, Nikolaos Louros, Elien Heylen, Kaat De Cremer, Katrijn De Brucker, Stijn Vereecke, Benno Verbelen, Pieter Vermeersch, Joost Schymkowitz, Frederic Rousseau, David Cassiman, Sarah-Maria Fendt, Arnout Voet, Bruno P A Cammue, Karin Thevissen, Kim De Keersmaecker
ABSTRACT
Metabolic rewiring is a hallmark of cancer that supports tumor growth, survival, and chemotherapy resistance. Although normal cells often rely on extracellular serine and glycine supply, a significant subset of cancers becomes addicted to intracellular serine/glycine synthesis, offering an attractive drug target. Previously developed inhibitors of serine/glycine synthesis enzymes did not reach clinical trials due to unfavorable pharmacokinetic profiles, implying that further efforts to identify clinically applicable drugs targeting this pathway are required. In this study, we aimed to develop therapies that can rapidly enter the clinical practice by focusing on drug repurposing, as their safety and cost-effectiveness have been optimized before. Using a yeast model system, we repurposed two compounds, sertraline and thimerosal, for their selective toxicity against serine/glycine synthesis-addicted breast cancer and T-cell acute lymphoblastic leukemia cell lines. Isotope tracer metabolomics, computational docking, enzymatic assays, and drug-target interaction studies revealed that sertraline and thimerosal inhibit serine/glycine synthesis enzymes serine hydroxymethyltransferase and phosphoglycerate dehydrogenase, respectively. In addition, we demonstrated that sertraline's antiproliferative activity was further aggravated by mitochondrial inhibitors, such as the antimalarial artemether, by causing G1-S cell-cycle arrest. Most notably, this combination also resulted in serine-selective antitumor activity in breast cancer mouse xenografts. Collectively, this study provides molecular insights into the repurposed mode-of-action of the antidepressant sertraline and allows to delineate a hitherto unidentified group of cancers being particularly sensitive to treatment with sertraline. Furthermore, we highlight the simultaneous inhibition of serine/glycine synthesis and mitochondrial metabolism as a novel treatment strategy for serine/glycine synthesis-addicted cancers.
Sigma-Aldrich
Benzalkonium chloride, ≥95.0% ((calculated on dry substance), T)
Sigma-Aldrich
NCT-503, ≥98% (HPLC)
Sigma-Aldrich
Sertraline hydrochloride, ≥98% (HPLC)
TPP® tissue culture plates, 96 well plate, flat bottom, polystyrene, 0.34cm2, sterile, 108/cs
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
SHMT inhibitor Hit 1, ≥98% (HPLC)