Merck
  • Retinoic Acid Synthesis Deficiency Fosters the Generation of Polymorphonuclear Myeloid-Derived Suppressor Cells in Colorectal Cancer.
Retinoic Acid Synthesis Deficiency Fosters the Generation of Polymorphonuclear Myeloid-Derived Suppressor Cells in Colorectal Cancer.
Cancer immunology research (2020-11-13)
Hong-Wei Sun, Jing Chen, Wen-Chao Wu, Yan-Yan Yang, Yi-Tuo Xu, Xing-Juan Yu, Hai-Tian Chen, Zilian Wang, Xiao-Jun Wu, Limin Zheng
ABSTRACT
Metabolism is reprogrammed in cancer to fulfill the demands of malignant cells for cancer initiation and progression. Apart from its effects within cancer cells, little is known about whether and how reprogramed metabolism regulates the surrounding tumor microenvironment (TME). Myeloid-derived suppressor cells (MDSC) are key regulators of the TME and greatly affect tumor progression and therapeutic responses. In this study, our results revealed that retinol metabolism-related genes and enzymes were significantly downregulated in human colorectal cancer compared with adjacent colonic tissues, and tumors exhibited a defect in retinoic acid (RA) synthesis. Reduced ADH1-mediated retinol metabolism was associated with attenuated RA signaling and accumulated MDSCs in colorectal cancer tumors. Using an in vitro model, generating MDSCs from CD34+ myeloid precursors, we found that exogenous RA could abrogate the generation of polymorphonuclear MDSCs (PMN-MDSC) with negligible impact on myeloid differentiation. Mechanistically, RA could restrain the glycolytic capacity of myeloid cells, which in turn activated the AMP-activated protein kinase (AMPK) pathway, further impairing the suppressive capacity of myeloid cells. Supplementation with RA could significantly delay tumor growth, with reduced arginase-1-expressing myeloid cells and increased CD8+ and granzyme B+ T cells in both colitis-associated and implanted MC38 mouse colorectal cancer models. Our results indicated that the defect in ADH1-mediated RA synthesis could provide a possible mechanism that fosters the generation of PMN-MDSCs in colorectal cancer and that restoring RA signaling in the TME could serve as a promising therapeutic strategy to abrogate the generation of PMN-MDSCs.
Sigma-Aldrich
Citrate Buffer, pH 6.0, 10×, Antigen Retriever, for immunohistochemistry
Pricing and availability is not currently available.
Sigma-Aldrich
EDTA Buffer, pH 8.5, 10×, Antigen Retriever
Pricing and availability is not currently available.
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%
Pricing and availability is not currently available.