Merck
  • LncRNA NEAT1 regulates chondrocyte proliferation and apoptosis via targeting miR-543/PLA2G4A axis.
LncRNA NEAT1 regulates chondrocyte proliferation and apoptosis via targeting miR-543/PLA2G4A axis.
Human cell (2020-10-12)
Peng Xiao, Xu Zhu, Jinpeng Sun, Yuhang Zhang, Weijian Qiu, Jianqiang Li, Xuejian Wu
ABSTRACT
Osteoarthritis (OA), which is characterized by articular cartilage degeneration, shows a gradually increasing incidence with age. This study explored the molecular mechanism underlying the proliferation and apoptosis of chondrocytes during OA progression. In this study, chondrocytes were isolated from human knee cartilages. The targeted relationship among nuclear enriched abundant transcript 1 (NEAT1), microRNA-543 (miR-543) and PLA2G4A was predicted on TargetScan V7.2 and starBase and validated by performing dual-luciferase reporter assay. High-expressed NEAT1 was detected in OA cartilage and chondrocytes. NEAT1 was negatively correlated with miR-543 and was low-expressed in OA cartilage and PLA2G4A was negatively correlated with miR-543 and was high-expressed in OA cartilage. In OA chondrocytes, the overexpressed NEAT1 inhibited the expressions of p-Akt1 and Bcl-2 and upregulated that of matrix metalloprotease (MMP)-3, MMP-9, MMP-13, interleukin (IL)-6 and IL-8, but such effects of overexpressed NEAT1 were reversed by miR-543 mimic. SiRNA-NEAT1 exerted an opposite effect to NEAT1 overexpression on OA chondrocytes, but this could be reversed by miR-543 inhibitor. The effect of PLA2G4A overexpression was the opposite to miR-543 mimic on OA chondrocytes. In conclusion, NEAT1 could sponge miR-543 to induce PLA2G4A expression, inhibit chondrocyte proliferation and promote apoptosis.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Hanks′ Balanced Salt solution, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Hematoxylin, certified by the BSC