Merck
  • A Disorder-to-Order Transition Activates an ATP-Independent Membrane Protein Chaperone.
A Disorder-to-Order Transition Activates an ATP-Independent Membrane Protein Chaperone.
Journal of molecular biology (2020-11-15)
Alex Siegel, Camille Z McAvoy, Vinh Lam, Fu-Cheng Liang, Gerard Kroon, Emily Miaou, Patrick Griffin, Peter E Wright, Shu-Ou Shan
ABSTRACT
The 43 kDa subunit of the chloroplast signal recognition particle, cpSRP43, is an ATP-independent chaperone essential for the biogenesis of the light harvesting chlorophyll-binding proteins (LHCP), the most abundant membrane protein family on earth. cpSRP43 is activated by a stromal factor, cpSRP54, to more effectively capture and solubilize LHCPs. The molecular mechanism underlying this chaperone activation is unclear. Here, a combination of hydrogen-deuterium exchange, electron paramagnetic resonance, and NMR spectroscopy experiments reveal that a disorder-to-order transition of the ankyrin repeat motifs in the substrate binding domain of cpSRP43 drives its activation. An analogous coil-to-helix transition in the bridging helix, which connects the ankyrin repeat motifs to the cpSRP54 binding site in the second chromodomain, mediates long-range allosteric communication of cpSRP43 with its activating binding partner. Our results provide a molecular model to explain how the conformational dynamics of cpSRP43 enables regulation of its chaperone activity and suggest a general mechanism by which ATP-independent chaperones with cooperatively folding domains can be regulated.
Sigma-Aldrich
Bistrifluoroacetamide
Sigma-Aldrich
DL-Cysteine, technical grade