Merck
Rational design of indoleamine 2,3-dioxygenase inhibitors.
Journal of medicinal chemistry (2010-01-09)
Ute F Röhrig, Loay Awad, Aurélien Grosdidier, Pierre Larrieu, Vincent Stroobant, Didier Colau, Vincenzo Cerundolo, Andrew J G Simpson, Pierre Vogel, Benoît J Van den Eynde, Vincent Zoete, Olivier Michielin
ABSTRACT
Indoleamine 2,3-dioxygenase (IDO) is an important therapeutic target for the treatment of diseases such as cancer that involve pathological immune escape. We have used the evolutionary docking algorithm EADock to design new inhibitors of this enzyme. First, we investigated the modes of binding of all known IDO inhibitors. On the basis of the observed docked conformations, we developed a pharmacophore model, which was then used to devise new compounds to be tested for IDO inhibition. We also used a fragment-based approach to design and to optimize small organic molecule inhibitors. Both approaches yielded several new low-molecular weight inhibitor scaffolds, the most active being of nanomolar potency in an enzymatic assay. Cellular assays confirmed the potential biological relevance of four different scaffolds.
Product Description
Supelco
8-Quinolinol, PESTANAL®, analytical standard
Sigma-Aldrich
Bis(5-chloro-2-hydroxyphenyl)methane, 95%
Sigma-Aldrich
8-Quinolinol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
N-Phenyl-p-phenylenediamine, 98%
Sigma-Aldrich
4-Hydroxycarbazole, 95%
Sigma-Aldrich
9-Fluorenone, 98%
Sigma-Aldrich
9-Hydroxyfluorene, 96%
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
8-Hydroxyquinoline, ACS reagent, 98.5%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Sigma-Aldrich
4-Amino-1-naphthol hydrochloride, technical grade, 90%
Sigma-Aldrich
8-Hydroxyquinoline