Merck
  • Cranial Suture Regeneration Mitigates Skull and Neurocognitive Defects in Craniosynostosis.
Cranial Suture Regeneration Mitigates Skull and Neurocognitive Defects in Craniosynostosis.
Cell (2021-01-09)
Mengfei Yu, Li Ma, Yuan Yuan, Xin Ye, Axel Montagne, Jinzhi He, Thach-Vu Ho, Yingxi Wu, Zhen Zhao, Naomi Sta Maria, Russell Jacobs, Mark Urata, Huiming Wang, Berislav V Zlokovic, Jian-Fu Chen, Yang Chai
ABSTRACT
Craniosynostosis results from premature fusion of the cranial suture(s), which contain mesenchymal stem cells (MSCs) that are crucial for calvarial expansion in coordination with brain growth. Infants with craniosynostosis have skull dysmorphology, increased intracranial pressure, and complications such as neurocognitive impairment that compromise quality of life. Animal models recapitulating these phenotypes are lacking, hampering development of urgently needed innovative therapies. Here, we show that Twist1+/- mice with craniosynostosis have increased intracranial pressure and neurocognitive behavioral abnormalities, recapitulating features of human Saethre-Chotzen syndrome. Using a biodegradable material combined with MSCs, we successfully regenerated a functional cranial suture that corrects skull deformity, normalizes intracranial pressure, and rescues neurocognitive behavior deficits. The regenerated suture creates a niche into which endogenous MSCs migrated, sustaining calvarial bone homeostasis and repair. MSC-based cranial suture regeneration offers a paradigm shift in treatment to reverse skull and neurocognitive abnormalities in this devastating disease.
Product Description
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
LY2090314, ≥98% (HPLC)
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Tamoxifen, ≥99%