Merck
  • New approach for drug susceptibility testing: monitoring the stress response of mycobacteria.
New approach for drug susceptibility testing: monitoring the stress response of mycobacteria.
Antimicrobial agents and chemotherapy (2009-08-26)
Ronald J Rieder, Zhihui Zhao, Boris Zavizion
ABSTRACT
Methods currently used for in vitro drug susceptibility testing are based on the assessment of bacterial growth-related processes. This reliance on cellular reproduction leads to prolonged incubation times, particularly for slowly growing organisms such as mycobacteria. A new rapid phenotypic method for the drug susceptibility testing of mycobacteria is described. The method is based on the detection of the physiological stress developed by susceptible mycobacterial cells in the presence of an antimicrobial compound. The induced stress was quantified by differential monitoring of the dielectric properties of the bacterial suspension, an easily measurable electronic property. The data presented here characterize the stress developed by Mycobacterium tuberculosis cells treated with rifampin (rifampicin), isoniazid, ethambutol, and pyrazinamide. Changes in the dielectric-based profiles of the drug-treated bacteria revealed the respective susceptibilities in near real time, and the susceptibilities were well correlated with conventional susceptibility test data.
Product Description
Millipore
Middlebrook 7H10 Agar Base, suitable for microbiology, NutriSelect® Plus, for use in isolation and cultivation of Mycobacterium species
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Pyrazinecarboxamide
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Millipore
Middlebrook 7H9 Broth Base, suitable for microbiology, NutriSelect® Plus,
Recommended for use in isolation and cultivation of Mycobacterium species