Merck
  • Targeting GM-CSF for collagenase-induced osteoarthritis pain and disease in mice.
Targeting GM-CSF for collagenase-induced osteoarthritis pain and disease in mice.
Osteoarthritis and cartilage (2020-02-07)
K M-C Lee, V Prasad, A Achuthan, A J Fleetwood, J A Hamilton, A D Cook
ABSTRACT
Pharmacological options for treating osteoarthritis (OA) are limited and alternative treatments are required. Given the clinical data indicating that granulocyte macrophage-colony stimulating factor (GM-CSF) may be a therapeutic target in human OA, we evaluated different treatment regimens with a neutralizing anti-GM-CSF monoclonal antibody (mAb) in an experimental OA model to determine their effectiveness on amelioration of pain and disease. The collagenase-induced osteoarthritis (CiOA) model was induced in C57BL/6 mice, followed by different treatment regimens of anti-GM-CSF mAb or isotype control. Anti-CCL17 mAb treatment was also administered continually during the late stage of CiOA. Pain-related behavior (change in weight distribution of hind limbs), and disease (cartilage damage and osteophyte size) were assessed. Blocking GM-CSF only during early synovitis in CiOA prevented pain and disease development. Once OA pain was established, regardless of the treatment regimen, anti-GM-CSF mAb treatment rapidly and efficiently ameliorated it; however, unless the treatment was continued, pain returned and disease progressed. Continual late stage blockade of GM-CSF was able to ameliorate pain (between-group difference: -6.567; 95% confidence interval (CI): -10.12, -3.011) and suppress cartilage damage (P = 0.0317, 95% CI: -1.75, -0.0556). Continual late stage blockade of CCL17 showed similar effects on pain and disease development. Early and short-term GM-CSF neutralization is effective at preventing CiOA pain and disease development but, once pain is evident, continual GM-CSF blockade is required to prevent pain from returning and to suppress disease progression in mice. These data reinforce the potential benefits of anti-GM-CSF (and anti-CCL17) mAb therapy in OA and should inform further clinical trials.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
Pricing and availability is not currently available.